Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Bovine Rotavirus Antigen ELISA Kit (Colorimetric)
SDP

Catalog No. nb601301kt
Click to view available options
Quantity:
1 Kit

High sensitivity and specificity for detection of bovine rotavirus antigen.

The Bovine Rotavirus Antigen ELISA Kit (Colorimetric) measures Bovine Rotavirus Antigen in feces.

Specifications

Assay Range Qualitative
Product Type ELISA Kit (Colorimetric)
Sample Type Fecal Extract
Specificity For detection of bovine rotavirus antigen
Target Species Bovine
For Use With (Application) ELISA
Kit Contents Instruction manual, Positive Control, Negative Control, Stop Solution, Wash Buffer, Substrate B, Substrate A, HRP-conjugate, Assay plate, Adhesive Strip (For 96 wells)
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Cell Biology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Rotavirus Antigen
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.