Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine RSV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA123354
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA123354 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA123354 Supplier Invitrogen™ Supplier No. PA123354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-23354 detects Bovine Respiratory Syncytial Virus in Virus samples.

Bovine Respiratory Syncytial Virus (BRSV) is a major cause of respiratory disease in young calves, contributing significantly to the bovine respiratory disease complex. As an enveloped virus belonging to the pneumovirus genus within the Paramyxoviridae family, BRSV contains a negative-sense, single-stranded RNA genome encoding 11 proteins. The virus infects both the upper and lower respiratory tracts and is primarily transmitted through nasal secretions. Closely related to the human respiratory syncytial virus (HRSV), BRSV shares similar epidemiological and pathogenic characteristics, making it a valuable model for studying respiratory infections. BRSV infection can produce symptoms ranging from mild respiratory distress to severe pneumonia, often exacerbated by secondary bacterial infections. The virus is notorious for its ability to evade the immune system by modulating immune responses, such as inhibiting effective CD8+ T cell responses, leading to chronic and severe respiratory complications in affected cattle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bovine RSV
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias BRSV
Host Species Rabbit
Immunogen Bovine Respiratory Syncytial Virus (BRSV) Fusion protein of the RB94 strain.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.