Learn More
Invitrogen™ Bovine RSV Polyclonal Antibody

Description
PA1-23355 detects Bovine Respiratory Syncytial Virus in Virus samples.
Specifications
Specifications
| Antigen | Bovine RSV |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | BRSV |
| Host Species | Rabbit |
| Immunogen | Human Respiratory Syncytial Virus (RSV) and Bovine Respiratory Syncytial Virus (BRSV) Fusion protein. |
| Purification Method | Affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.