Learn More
Invitrogen™ Bovine RSV strain RB94 Polyclonal Antibody

Description
PA1-4103 detects the bovine respiratory syncytial virus strain RB94. PA1-4103 has been successfully used in Western blot procedures. The PA1-4103 immunogen is a synthetic peptide corresponding to residues K(201) E L L P K V N N H D(211) C of bovine RSV strain (RB94).
Specifications
Specifications
| Antigen | Bovine RSV strain RB94 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | BRSV |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to residues K(201) E L L P K V N N H D(211) C of bovine RSV (strain RB94). |
| Purification Method | Antigen affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.