Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bovine Serum Albumin Goat anti-Bovine, Biotin, Polyclonal, Southern Biotech™

Goat Polyclonal Antibody
Supplier: Southern Biotechnology Associates 207908
Description
SouthernBiotech offers serum albumin antibodies for ELISA applications.
Specifications
Bovine Serum Albumin | |
Polyclonal | |
Biotin | |
Goat | |
Affinity Purified | |
RUO | |
Bovine | |
IgG |
ELISA | |
0.5mg/mL | |
Supplied in PBS/NaN3 | |
Bovine serum albumin | |
1.0 mg | |
Primary | |
Store at 2-8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction