Learn More
Invitrogen™ Bovine Serum Albumin Polyclonal Antibody, FITC

Description
PA1-29252 detects Bovine Serum Albumin from bovine samples.
Specifications
Specifications
| Antigen | Bovine Serum Albumin |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with proprietary stabilizer and 0.1% sodium azide |
| Gene | ALB |
| Gene Accession No. | P02769 |
| Gene Alias | ALB; ALB protein; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; Bos d 6; bovine albumin; bovine serum albumin; BSA; cell growth inhibiting protein 42; FDAH; GIG20; GIG42; growth-inhibiting protein 20; MGC179277; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 |
| Gene Symbols | ALB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.