Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bovine TNF-alpha Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2279
This item is not returnable.
View return policy
Description
TNF-alpha Polyclonal specifically detects TNF-alpha in Bovine samples. It is validated for Western Blot, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
TNF-alpha | |
Polyclonal | |
Unconjugated | |
Q06599 | |
TNF | |
E. coli-derived recombinant bovine TNF-alpha Leu78-Leu234 Accession # Q06599 | |
100 μg | |
Primary | |
Detects bovine TNF-alpha in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant human TNF-alpha , recombinant mouse TNF-alpha , recombinant rat TNF-alpha , recombinant feline TNF-alpha , recombinant equine TNF-alpha , recombinant porcine TNF-alpha , recombinant cotton rat TNF-alpha , and recombinant rhesus macaque TNF-alpha is observed. | |
Bovine | |
IgG |
Western Blot, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Goat | |
Affinity Purified | |
RUO | |
7124 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction