Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Bovine VEGF 164 Protein

Catalog No. 8917BV025CF
Click to view available options
Quantity:
25 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured in a cell proliferation assay using bAEC bovine aortic endothelial cells. The ED50 for this effect is 0.3-1.8 ng/mL.

Specifications

Accession Number P15691
For Use With (Application) Bioactivity
Gene ID (Entrez) 7422
Molecular Weight (g/mol) Observed M.W.: 21-26kDa, reducing conditions, Theoretical M.W.: 19kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived bovine VEGF protein Ala27-Arg190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Common Name VEGF
Gene Symbol VEGFA
Biological Activity Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is typically 0.3-1.8ng/mL.
Conjugate Unconjugated
Species Bovine
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ala27-Arg190
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.