Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BPGM Antibody (2E11), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259493APC
This item is not returnable.
View return policy
Description
BPGM Monoclonal antibody specifically detects BPGM in Human samples. It is validated for Flow CytometrySpecifications
BPGM | |
Monoclonal | |
APC | |
2,3-bisphosphoglycerate mutase, bisphosphoglycerate mutase, BPG-dependent PGAM, EC 3.1.3.132,3-bisphosphoglycerate synthase, EC 5.4.2.1, EC 5.4.2.4, erythrocyte 2,3-bisphosphoglycerate mutase2,3-bisphosphoglycerate mutase, erythrocyte | |
Recombinant human BPGM (1-259aa) purified from E. coli. | |
0.1 mL | |
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
669 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
200000000000 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction