Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPHL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580382
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580382 Supplier Invitrogen™ Supplier No. PA580382
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human BPHL.

Serine hydrolase that catalyzes the hydrolytic activation of amino acid ester prodrugs of nucleoside analogs such as valacyclovir and valganciclovir. Activates valacyclovir to acyclovir. May play a role in detoxification processes. It is a specific alpha-amino acid ester hydrolase that prefers small, hydrophobic, and aromatic side chains and does not have a stringent requirement for the leaving group other than preferring a primary alcohol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPHL
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene BPHL
Gene Accession No. Q86WA6
Gene Alias 2010012D11Rik; 5730533B08Rik; AI115341; biphenyl hydrolase-like (serine hydrolase); biphenyl hydrolase-like (serine hydrolase, breast epithelial mucin-associated antigen); biphenyl hydrolase-like protein; Biphenyl hydrolase-related protein; BPHL; BPH-RP; Breast epithelial mucin-associated antigen; MCNAA; PARG; Poly(ADP-ribose) glycohydrolase; VACVASE; valacyclovir hydrolase; valacyclovirase
Gene Symbols BPHL
Host Species Rabbit
Immunogen Recombinant Human BPHL protein (Ser38-Gln291).
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 670
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.