Learn More
BPI Antibody (971526), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB74681MFV610
Description
BPI Monoclonal antibody specifically detects BPI in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow CytometrySpecifications
| BPI | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry, Flow Cytometry | |
| 971526 | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry | |
| bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, CAP 57 | |
| Human embryonic kidney cell line HEK-293-derived transfected with human BPI, Val32-Lys487, Accession # P17213 | |
| RUO | |
| 671 | |
| Store at 4C in the dark. |