Learn More
Invitrogen™ BPI Monoclonal Antibody (4H5)

Description
MA1-40128 detects BPI from human samples.The MA1-40128 immunogen is full length human BPI. MA1-40128 recognizes only free BPI and does not interact with BPI that has formed a complex with LPS. MA1-40128 has been successfully used in ELISA applications. ELISA: monoclonal antibody 4H5 can be used both as coating and as detector.
Specifications
Specifications
| Antigen | BPI |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4H5 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | BPI |
| Gene Accession No. | P17213 |
| Gene Alias | 9230105K17Rik; Bactericidal permeability-increasing protein; bactericidal permeablility increasing protein; bactericidal/permeability-increasing protein; BPI; BPI fold containing family D, member 1; BPIFD1; CAP 57; rBPI; recombinant BPI holoprotein, rBPI; RP13-218F6_A.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.