Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BPI Polyclonal Antibody
BPI Polyclonal Antibody
Supplier: Thermo Scientific PA563942
Description
BPI Polyclonal Antibody for IHC

Specifications
BPI | |
Polyclonal | |
Unconjugated | |
BPI | |
9230105K17Rik; Bactericidal permeability-increasing protein; bactericidal permeablility increasing protein; bactericidal/permeability-increasing protein; BPI; BPI fold containing family D, member 1; BPIFD1; CAP 57; rBPI; recombinant BPI holoprotein, rBPI; RP13-218F6_A.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
671 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P17213 | |
BPI | |
Recombinant protein corresponding to Human BPI. Recombinant protein control fragment (Product #RP-103394). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction