Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BPIFB1 Antibody (086), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290203PCP
Description
BPIFB1 Monoclonal antibody specifically detects BPIFB1 in Human samples. It is validated for ELISASpecifications
BPIFB1 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
92747 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
086 | |
ELISA | |
bA49G10.6, chromosome 20 open reading frame 114, dJ1187J4.1, long palate, lung and nasal epithelium carcinoma associated 1, long palate, lung and nasal epithelium carcinoma-associated protein 1, LPLUNC1, MGC14597, VEMSGP, Von Ebner minor salivary gland protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human BPIFB1 (Accession#: AAH08429.1; Met1-Gln484). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction