Learn More
Invitrogen™ BPIFB3 Polyclonal Antibody

Description
The antibody detects endogenous level of total BPIFB3 protein.
Specifications
Specifications
| Antigen | BPIFB3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | BPIFB3 |
| Gene Accession No. | P59826 |
| Gene Alias | antimicrobial peptide RYA3; BPI fold containing family B member 3; BPI fold containing family B, member 3; BPI fold-containing family B member 3; Bpifb3; C20orf185; Ligand-binding protein RYA3; long palate, lung and nasal epithelium carcinoma-associated protein 3; Lplunc3; RYA3 |
| Gene Symbols | BPIFB3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.