Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPIFB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561086 Supplier Invitrogen™ Supplier No. PA561086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSLGALGSVE FSLATLPLIS NQYIELDINP IVKSVAGDII DFPKSRAPAK VPPKKDHTSQ VMVPLYLFNT TFGL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

BPI fold-containing family B member 3 may have the capacity to recognize and bind specific classes of odorants. May act as a carrier molecule, transporting odorants across the mucus layer to access receptor sites. May serve as a primary defense mechanism by recognizing and removing potentially harmful odorants or pathogenic microorganisms from the mucosa or clearing excess odorant from mucus to enable new odorant stimuli to be received.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPIFB3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BPIFB3
Gene Accession No. P59826
Gene Alias antimicrobial peptide RYA3; BPI fold containing family B member 3; BPI fold containing family B, member 3; BPI fold-containing family B member 3; Bpifb3; C20orf185; Ligand-binding protein RYA3; long palate, lung and nasal epithelium carcinoma-associated protein 3; Lplunc3; RYA3
Gene Symbols BPIFB3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BPIFB3. Recombinant protein control fragment (Product #RP-99465).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359710
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.