Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BPIFB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597518
Description
BPIFB3 Polyclonal Antibody for Western Blot, ELISA
BPI fold-containing family B member 3 may have the capacity to recognize and bind specific classes of odorants. May act as a carrier molecule, transporting odorants across the mucus layer to access receptor sites. May serve as a primary defense mechanism by recognizing and removing potentially harmful odorants or pathogenic microorganisms from the mucosa or clearing excess odorant from mucus to enable new odorant stimuli to be received.
Specifications
BPIFB3 | |
Polyclonal | |
Unconjugated | |
BPIFB3 | |
antimicrobial peptide RYA3; BPI fold containing family B member 3; BPI fold containing family B, member 3; BPI fold-containing family B member 3; Bpifb3; C20orf185; Ligand-binding protein RYA3; long palate, lung and nasal epithelium carcinoma-associated protein 3; Lplunc3; RYA3 | |
Rabbit | |
Protein G | |
RUO | |
359710, 378700 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.9 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P59826, Q80ZU7 | |
BPIFB3 | |
Recombinant Human BPI fold-containing family B member 3 protein (248-352AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction