Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPIFB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144817
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144817 Supplier Invitrogen™ Supplier No. PA5144817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PKDLETTICL IDVDTELLAS FSIEGDKLMI DAKLEKTSLN LRTSNVGNFD IGLMEVLVEK IFDLAFMPAM NAVLG Antigen sequence identity: Mouse 85%; Rat 85%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BPIFB4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BPIFB4
Gene Accession No. P59827
Gene Alias BPI fold containing family B member 4; BPI fold containing family B, member 4; BPI fold-containing family B member 4; BPIFB4; C20orf186; dJ726C3.5; Gm1006; LBP / BPI / CETP family, N-terminal domain containing protein; ligand-binding protein RY2G5; likely ortholog of rat probable ligand-binding protein RY2G5; long palate, lung and nasal epithelium carcinoma associated 4; long palate, lung and nasal epithelium carcinoma-associated protein 4; LPLUNC4; Novel LBP/BPI/CETP family, N-terminal domain containing protein; potential ligand-binding protein; RY2G5
Gene Symbols BPIFB4
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 511-585. Recombinant protein control fragment (Product #RP-109953).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.