Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPIL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563818 Supplier Invitrogen™ Supplier No. PA563818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVLPRHVGTE GSMATVGLSQ QLFDSALLLL QKAGALNLDI TGQLRSDDNL LNTSALGRLI PEVARQFPEP Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

BPIL1 is a member of the lipid transfer/lipopolysaccharide binding protein (LT/LBP) family. This protein is highly expressed in hypertrophic tonsils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPIL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BPIFB2
Gene Accession No. Q8N4F0
Gene Alias 2310034L21Rik; 2310069A01Rik; AV089125; Bactericidal/permeability-increasing protein-like 1; BPI fold containing family B member 2; BPI fold containing family B, member 2; BPI fold-containing family B member 2; Bpifb2; Bpil1; BPI-like 1; C20orf184; dJ726C3.2; Long palate, lung and nasal epithelium carcinoma-associated protein 2; LPLUNC2; RYSR; UNQ2489/PRO5776
Gene Symbols BPIFB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BPIL1. Recombinant protein control fragment (Product #RP-103367).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80341
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.