Learn More
Invitrogen™ BPIL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595697
Description
Immunogen sequence: IGKAPLQRAL QVTVPHFLDW SGEALQPTRI RILNVHVPRL HLKFIAGFGV RLLAAANFTF KVFRAPEPLE LTLPVELLAD TRVTQSSIRT PVVSISACSL FSGHANEFDG SNSTSHALLV LVQKHIKAVL SNKLCLSISN LVQGVNVHLG TLIGLNPVGP ESQIRYSMVS VPTVTSDYIS LEVNAVLFLL GKPIILPTDA TPFVLPRHVG TEGSMATVGL SQQLFDSALL LLQKAGALNL DITGQLRSDD NLLNTSALGR LIPEVARQFP EPMPVVLKVR LGATPVAMLH TNNATLRLQP F; Positive Samples: Mouse brain, Rat brain; Cellular Location: Secreted.
BPIL1 is a member of the lipid transfer/lipopolysaccharide binding protein (LT/LBP) family. This protein is highly expressed in hypertrophic tonsils.
Specifications
BPIL1 | |
Polyclonal | |
Unconjugated | |
BPIFB2 | |
2310034L21Rik; 2310069A01Rik; AV089125; Bactericidal/permeability-increasing protein-like 1; BPI fold containing family B member 2; BPI fold containing family B, member 2; BPI fold-containing family B member 2; Bpifb2; Bpil1; BPI-like 1; C20orf184; dJ726C3.2; Long palate, lung and nasal epithelium carcinoma-associated protein 2; LPLUNC2; RYSR; UNQ2489/PRO5776 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
296290, 66557 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.42 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8C1E1 | |
BPIFB2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 40-340 of human BPIFB2 (NP_0795031). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.