Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPIL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988551
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8551 Supplier Bioss Supplier No. BS8423R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BPIL1 is a member of the lipid transfer/lipopolysaccharide binding protein (LT/LBP) family. This protein is highly expressed in hypertrophic tonsils.

Specifications

Antigen BPIL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BPIFB2
Gene Accession No. Q8N4F0
Gene Alias 2310034L21Rik; 2310069A01Rik; AV089125; Bactericidal/permeability-increasing protein-like 1; BPI fold containing family B member 2; BPI fold containing family B, member 2; BPI fold-containing family B member 2; BPIFB2; BPIL1; BPI-like 1; C20orf184; dJ726C3.2; Long palate, lung and nasal epithelium carcinoma-associated protein 2; LPLUNC2; RYSR; UNQ2489/PRO5776
Gene Symbols BPIFB2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BPIL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80341
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.