Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPIL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560316 Supplier Invitrogen™ Supplier No. PA560316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLRFENILSS ILHFGVLPLA NAKLQQGFPL SNPHKFLFVN SDIEVLEGFL LISTDLKYET SSKQQPSFHV WEGLN Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 80%.

BPIFC gene ontology annotations related to this gene include lipopolysaccharide binding; phospholipid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPIL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BPIFC
Gene Accession No. Q8NFQ6
Gene Alias 4732454E24; bactericidal/permeability-increasing protein-like 2; BPI fold containing family C; BPI fold-containing family C protein; Bpifc; Bpil2; BPI-like 2
Gene Symbols BPIFC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BPIL2. Recombinant protein control fragment (Product #RP-102939).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254240
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.