Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BPIL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571441
Description
BPIL2 Polyclonal Antibody for Western Blot
BPIFC gene ontology annotations related to this gene include lipopolysaccharide binding; phospholipid binding.
Specifications
BPIL2 | |
Polyclonal | |
Unconjugated | |
BPIFC | |
4732454E24; bactericidal/permeability-increasing protein-like 2; BPI fold containing family C; BPI fold-containing family C protein; Bpifc; Bpil2; BPI-like 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
254240 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NFQ6 | |
BPIFC | |
synthetic peptide directed towards the C terminal region of human BPIL2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction