Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518787
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518787 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518787 Supplier Invitrogen™ Supplier No. PA518787
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

BPNT1, also called bisphosphate 3-prime-nucleotidase, or BPntase, is a member of a magnesium-dependent phosphomonoesterase family. Lithium, a major drug used to treat manic depression, acts as an uncompetitive inhibitor of BPntase. The predicted human protein is 92% identical to mouse BPntase. BPntase's physiologic role in nucleotide metabolism may be regulated by inositol signaling pathways. The inhibition of human BPntase may account for lithium-induced nephrotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPNT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Bpnt1
Gene Accession No. O95861
Gene Alias 3'(2'), 5'-bisphosphate nucleotidase 1; 3'(2'),5'-bisphosphate nucleotidase 1; Bisphosphate 3'-nucleotidase 1; BPNT1; BPntase; epididymis luminal protein 20; HEL20; PAP-inositol 1,4-phosphatase; PAP-inositol-1,4-phosphatase; PIP; Sal3; scHAL2 analogous 3; testicular tissue protein Li 29
Gene Symbols Bpnt1
Host Species Goat
Immunogen Synthetic peptide sequence (RNYDYYASRVPESIK) corresponding to the C-terminus amino acids of BPNT1 (aa 289-304).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10380
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.