Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPOZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120648 Supplier Invitrogen™ Supplier No. PA5120648
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse skeletal muscle, Rat skeletal muscle. Immunogen sequence: MDTSDLFASC RKGDVGRVRY LLEQRDVEVN VRDKWDSTPL YYACLCGHEE LVLYLLANGA RCEANTFDGE RCLYGALSDP IRRALRDYKQ VTASCRRRDY YDDFLQRLLE QGIHSDVVFV VHGKPFRVHR CVLGARSAYF.

This gene encodes a protein with an ankyrin repeat region and two BTB/POZ domains, which are thought to be involved in protein-protein interactions. Expression of this gene is activated by the phosphatase and tensin homolog, a tumor suppressor. Alternate splicing results in three transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPOZ
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ABTB1
Gene Accession No. Q5XIU1, Q99LJ2
Gene Alias ABTB1; abtb1 {ECO:0000312; AI847549; ankyrin repeat and BTB (POZ) domain containing 1; ankyrin repeat and BTB domain containing 1; ankyrin repeat and BTB/POZ domain-containing protein 1; BC003234; BPOZ; BTB3; BTBD21; EF1ABP; elongation factor 1A-binding protein; EMBL:AAH83579.1}; mFLJ00331 protein; MGC20585; PP2259
Gene Symbols ABTB1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human ABTB1 (NP_742024.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297432, 80283
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.