Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BPTES
GREENER_CHOICESDP

Catalog No. 530110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
530110 10 mg
530150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 530110 Supplier Tocris Bioscience™ Supplier No. 5301/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Glutaminase Inhibitors

BPTES is an allosteric glutaminase (GLS1) inhibitor (IC50 = 3.3 μM). Selective for GLS1 over GLS2 and γ-glutamyl transpeptidase. Induces cell death of P493 human lymphoma B cells in vitro and reduces tumor volume of P493 cell xenografts in mice.

Specifications

Chemical Name or Material N,N′-[Thiobis(2,1-ethanediyl-1,3,4-thiadiazole-5,2-diyl)]bisbenzeneacetamide
CAS 314045-39-1
Quantity 10 mg
Target Hydrolase Inhibitors
Molecular Formula C24H24N6O2S3
Formula Weight 524.68
Purity >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.