Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BPY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570656 Supplier Invitrogen™ Supplier No. PA570656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene is located in the nonrecombining portion of the Y chromosome, and expressed specifically in testis. The encoded protein interacts with ubiquitin protein ligase E3A and may be involved in male germ cell development and male infertility. Three nearly identical copies of this gene exist on chromosome Y; two copies are part of a palindromic region. This record represents the copy outside of the palidromic region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPY2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BPY2
Gene Accession No. O14599
Gene Alias Basic charge, Y-linked 2; basic charge, Y-linked, 2; basic charge, Y-linked, 2C; basic protein on Y chromosome 2; BPY2; BPY2A; BPY2B; BPY2C; testis-specific basic protein on Y, 2; testis-specific basic protein Y 2; variable charge, Y-linked, 2; variably charged protein Y 2; VCY2; VCY2A; VCY2B; VCY2C; y-linked
Gene Symbols BPY2, BPY2C
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human BPY2 (aa 57-106).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442868, 9083
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.