Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BPY2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12882R

Description
This gene is located in the nonrecombining portion of the Y chromosome, and expressed specifically in testis. The encoded protein interacts with ubiquitin protein ligase E3A and may be involved in male germ cell development and male infertility. Three nearly identical copies of this gene exist on chromosome Y; two copies are part of a palindromic region. This record represents the copy outside of the palidromic region.Specifications
BPY2 | |
Polyclonal | |
Unconjugated | |
BPY2C | |
Basic charge, Y-linked 2; basic charge, Y-linked, 2; basic charge, Y-linked, 2C; basic protein on Y chromosome 2; BPY2; BPY2A; BPY2B; BPY2C; testis-specific basic protein on Y, 2; Testis-specific basic protein Y 2; variable charge, Y-linked, 2; Variably charged protein Y 2; VCY2; VCY2A; VCY2B; VCY2C | |
Rabbit | |
Protein A | |
RUO | |
442868, 9083 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O14599 | |
BPY2, BPY2C | |
KLH conjugated synthetic peptide derived from human BPY2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction