Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BQ 788 sodium salt
GREENER_CHOICESDP

Catalog No. 15001R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-001-R 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-001-R Supplier Tocris Bioscience™ Supplier No. 1500/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective ETB antagonist

Potent, selective ETB receptor antagonist (IC50 = 1.2 nM for inhibition of ET-1 binding to human Girardi heart cells).

Chemical Identifiers

Molecular Formula C34H50N5NaO7

Specifications

Quantity 1 mg
Formula Weight 663.79
Physical Form Solid
Purity >98%
Chemical Name or Material BQ 788 sodium salt
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.