Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brachyury Antibody (6C12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Brachyury |
---|---|
Clone | 6C12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Description
Brachyury Monoclonal specifically detects Brachyury in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
Brachyury | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
RUO | |
Human | |
brachyury protein, MGC104817, Protein T, SAVA, T brachyury (mouse) homolog, T brachyury homolog, T Brachyury Transcription Factor, T, brachyury homolog (mouse), T-Box Transcription Factor T, TBXT, TF, TFT | |
T | |
IgG1 | |
Affinity Purified |
6C12 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Mouse | |
Hematopoietic Stem Cell Markers, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
PBS with 0.05% Sodium Azide | |
6862 | |
Purified recombinant fragment of human T (AA: 218-352) expressed in E. Coli. | |
Primary | |
47.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title