Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brachyury Monoclonal Antibody (07), Invitrogen™
Mouse Monoclonal Antibody
$296.50 - $511.50
Specifications
Antigen | Brachyury |
---|---|
Clone | 07 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543480
|
Invitrogen™
MA543480 |
50 μL |
Each for $296.50
|
|
|||||
PIMA543052
|
Invitrogen™
MA543052 |
100 μL |
Each for $511.50
|
|
|||||
Description
Specificity: Human Brachyury/TBXT
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.Specifications
Brachyury | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3 | |
TBXT | |
IgG2b | |
Protein A | |
Antibody |
07 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
O15178 | |
6862 | |
A synthetic peptide corresponding to the C-terminus of the Human Brachyury/TBXT. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
TBXT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title