Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Brachyury Monoclonal Antibody (X1AO2), eBioscience™

Catalog No. 14977037
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-977-037 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-977-037 Supplier Invitrogen™ Supplier No. 14977037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody X1AO2 recognizes human Brachyury, a 47 kDa protein encoded by the T gene and functioning as a nuclear transcription factor. This transcription factor binds to specific DNA regions near the palindromic sequence TCACACCT (T-site) through a region in its N-terminus, called the T-box. Brachyury functions during embryogenesis to regulate midline development by establishing the anterior/posterior axis through the regulation of genes involved in mesoderm formation and differentiation. Brachyury is the primary mesoderm marker used to distinguish differentiation of embryonic stem cells into a mesenchymal stem cell lineage. Variations in the T gene in humans are associated with neural tube defects and chordomas. Expression of brachyury is also increased in advanced stage lung tumors and tumor cell lines. Applications Reported: This X1AO2 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, immunohistochemical staining, and immunocytochemistry. Applications Tested: This X1AO2 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 5 μg/mL. This X1AO2 antibody has also been tested by immunocytochemistry of fixed and permeabilized human cells and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be...

The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brachyury
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone X1AO2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TBXT
Gene Accession No. O15178
Gene Alias Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3
Gene Symbols TBXT
Host Species Mouse
Immunogen E.coli-derived fragment of human Brachyury corresponding to amino acids 237-348.
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6862
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.