Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Brachyury Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582079 Supplier Invitrogen™ Supplier No. PA582079
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQPGYSQWGW LLPGTSTLCP PANPHPQFGG ALSLPSTHSC DRYPTLRSHR SSPYPSPYAH RNNSPTYSDN SPACLSMLQS HDNWSSLGMP AHPSMLPVSH NASPPTSSSQ Y Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 85%.

The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brachyury
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBXT
Gene Accession No. O15178
Gene Alias Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3
Gene Symbols TBXT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human T. Recombinant protein control fragment (Product #RP-100738).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6862
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.