Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poltex Bracket for Pipette Controller

The Poltex Bracket for Pipette Controller is ideal for hanging pipette controller securely up and out of the way.
Supplier: Poltex BRKT
Description
- Pipette Controller hole is 1.125 in. diameter
- PETG construction
- Available mounting options include VHB tape, powerful magnets, or keyhole slots for wall mount
Specifications
Not Autoclavable | |
Pipette Controller, Poltex Serological Pipette Holders | |
63.5 mm | |
114 mm |
PETG | |
VHB Tape | |
70 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction