Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biodegradable Flagging Tapes

Catalog No. p-6567168 Shop All Brady Corporation Products
Click to view available options
Color:
Black, Red
Black, Yellow
Displayed Text:
Caution
Danger

Catalog No. 19-104-304

Supplier: Brady™ 91088

Used when hazardous marking is required in ecology conscious situations

  • Made of 70% wood fiber
  • Non-toxic
  • Will not harm wildlife, livestock or the environment
  • Complete degradation will occur in 6-12 months depending on environment conditions

Specifications

Color Black, Yellow
Width (English) 3 in.
Width (Metric) 7.62 cm
Length (English) 150 ft.
Length (Metric) 45.7 m
Displayed Text Danger
Type Biodegradable Flagging Tape
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.