Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Biohazard with Pictogram Signs
Shop All Brady Corporation Products
Click to view available options
Height (English):
3.5 in.
5 in.
7 in.
Width (English):
3.5 in.
5 in.
10 in.
Description
Bright colors and bold text provide high visibility to critical messages.
- Biohazard marking is required by safety code 1910.1030(g)(1)
- Designed to withstand harsh industrial environments
- Black on orange
- B-485 self-sticking polyester — polyester film with sensitive acrylic
Compliance
Meets OSHA Standards 1910.145(e)(4) and 1910.1030(g)(1)
Specifications
Specifications
Material | Polyester |
Description | High Performance Polyester |
Height (English) | 3.5 in. |
Height (Metric) | 8.89 cm |
Width (English) | 5 in. |
Width (Metric) | 12.7 cm |
Dimensions (L x W) | 3.5 x 5 in. |
Displayed Text | BIOHAZARD |
Product Type | Biohazard Sign with Pictogram |
Color | Black on Orange |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction