Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Biohazard with Pictogram Signs

Catalog No. p-3697618
Click to view available options
Height (English):
3.5 in.
5 in.
7 in.
Width (English):
3.5 in.
5 in.
10 in.

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

Bright colors and bold text provide high visibility to critical messages.

 

  • Biohazard marking is required by safety code 1910.1030(g)(1)
  • Designed to withstand harsh industrial environments
  • Black on orange
  • B-485 self-sticking polyester — polyester film with sensitive acrylic

 

Compliance

Meets OSHA Standards 1910.145(e)(4) and 1910.1030(g)(1)

Specifications

Material Polyester
Description High Performance Polyester
Height (English) 3.5 in.
Height (Metric) 8.89 cm
Width (English) 5 in.
Width (Metric) 12.7 cm
Dimensions (L x W) 3.5 x 5 in.
Displayed Text BIOHAZARD
Product Type Biohazard Sign with Pictogram
Color Black on Orange

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.