Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Bradylink™ Solid Color Warning Posts

Catalog No. 19104355
Click to view available options
Color:
Black
White
Yellow
Packaging Quantity:
1 Ea.
6/Pk.
Height (English):
40 in.
41 in.

Provides sturdy support for barricading chain and tapes

  • Posts are non-conductive so can be safely used in electrically hazardous areas
  • Hollow base can be filled with sand for more stable application
  • Large C-hook holds chain in place on large posts
  • Can be used open or closed for greater security

Specifications

Color Yellow
Diameter (English) Base 14 in.
Packaging Quantity 6/Pk.
Diameter (English) Outer 2.5 in.
Diameter (Metric) Base 35.5 cm
Diameter (Metric) Outer 6.35 cm
Height (English) 40 in.
Height (Metric) 101.6 cm
Material Polyethylene, Polystyrene
Type Warning Post
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.