Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Bradylink™ Solid Color Warning Posts
Click to view available options
Color:
Black
White
Yellow
Packaging Quantity:
1 Ea.
6/Pk.
Height (English):
40 in.
41 in.
Description
- Posts are non-conductive so can be safely used in electrically hazardous areas
- Hollow base can be filled with sand for more stable application
- Large C-hook holds chain in place on large posts
- Can be used open or closed for greater security
Specifications
Specifications
Color | Yellow |
Height (English) | 40 in. |
Packaging Quantity | 1 Ea. |
Height (Metric) | 101.6 cm |
Type | Warning Post |
Material | Polyethylene, Polystyrene |
Diameter (Metric) Base | 35.5 cm |
Diameter (Metric) Outer | 6.35 cm |
Diameter (English) Outer | 2.5 in. |
Diameter (English) Base | 14 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction