Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Caution and Warning Tapes

Catalog No. 19807082 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black on White
Black on Yellow
Magenta on Yellow
Red on White
White
White Photoluminescent
Yellow
Dimensions (L x W):
15 ft. x 2 in.
15 ft. x 3 in.
5 yd. x 2 in.
54 ft. x 1 in.
54 ft. x 2 in.
54 ft. x 3 in.
Material:
Encapsulated Vinyl
Reflective Vinyl
Vinyl
Description:
Checkered
Solid
Stripes
Catalog No. Color Dimensions (L x W) Material Description
19-807-082 Magenta on Yellow 54 ft. x 3 in. Encapsulated Vinyl Stripes
19-016186 Red on White 54 ft. x 1 in. Encapsulated Vinyl Stripes
19-016190 Black on Yellow 54 ft. x 2 in. Encapsulated Vinyl Stripes
19-016185 Black on White 54 ft. x 2 in. Encapsulated Vinyl Stripes
19-016191 Black on Yellow 54 ft. x 3 in. Encapsulated Vinyl Stripes
19-017131 Black on Yellow 15 ft. x 3 in. Reflective Vinyl Stripes
19-017257 White 15 ft. x 2 in. Reflective Vinyl Solid
19-017258 Yellow - Reflective Vinyl Solid
19-017259 White 15 ft. x 3 in. Reflective Vinyl Solid
19-807-242 Black on White 54 ft. x 2 in. Encapsulated Vinyl Checkered
19-807-240 Black on White - Reflective Vinyl Stripes
19-807-241 Red on White - Reflective Vinyl Stripes
19-807-243 Black on Yellow 54 ft. x 2 in. Encapsulated Vinyl Checkered
19-807-257 Yellow 15 ft. x 3 in. Reflective Vinyl Solid
19-807-244 White Photoluminescent 5 yd. x 2 in. Vinyl Solid
15 options

Catalog No. 19-807-082

Supplier: Brady™ 55314

Only null left
Add to Cart
Add to Cart

Various tape designs in choice of two materials

  • Ideal for temporary use outdoors or permanent indoor warning
  • Sold as roll

B-957 Reflective Vinyl with Adhesive

  • Thickness: 1.65 to 1.91mm (0.065 to 0.075 in.)
  • Service temp.: -40° to 70°C (-40° to 160°F)

B-950 Encapsulated Vinyl with Adhesive

  • Thickness: 0.15mm (0.006 in.)
  • Service temp.: -40° to 80°C (-40° to 176°F)

Compliance

B-950 warning stripe and check tape meets OSHA specifications.

Specifications

Dimensions (L x W) 54 ft. x 3 in.
Description Stripes
Adhesive Type Rubber-based Pressure-Sensitive
Color Magenta on Yellow
For Use With (Application) Marking physical hazards
Width (English) 3 in.
Width (Metric) 7.62 cm
Temperature Range (English) -40°F to +176°F
Temperature Range (Metric) -40°C to +80°C
Thickness 0.15 mm
Length (Metric) 16.45 m
Length (English) 54 ft.
Certifications/Compliance OSHA 1910.144
Material Encapsulated Vinyl
Type Warning Stripe and Check Tape
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.