Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Chemical and Hazardous Materials Signs: Diesel Fuel

Catalog No. p-3697699 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
14 x 10 in.
Material:
Fiberglass
Polyester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Material
19-103-653 14 x 10 in. Polyester
19-016610 - Fiberglass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-103-653 Supplier Brady™ Supplier No. 84376
Only null left
Add to Cart
Add to Cart

For identifying plant's chemical storage and hazardous materials area

  • Keep employees safe by indicating how chemical are stored
  • With OSHA standards
  • Available in variety of materials and sizes including photoluminescent

Specifications

Material Polyester
Quantity 1 Ea.
Description Self-sticking Polyester
Height (English) 10 in.
Width (English) 14 in.
Dimensions (L x W) 14 x 10 in.
Displayed Text DANGER - DIESEL FUEL
Product Type Chemical and Hazardous Materials Sign
Color Red on White

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.