Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Danger: Hazardous Waste Storage Area… Signs

Catalog No. 19035596 Shop All Brady Corporation Products
Click to view available options
Material:
Fiberglass
Plastic
Polyester
Width (English):
10 in.
14 in.

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Sign reads “DANGER HAZARDOUS WASTE STORAGE AREA UNAUTHORIZED PERSONS KEEP OUT”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
  • B-555
    • Aluminum — 0.040 in. coil coated aluminum construction

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145, ASTM D 1593
    Material Plastic
    Quantity 1 Ea.
    Mounting Type Surface Mount
    For Use With (Application) Light duty industrial, utility, commercial, institutional environments
    Height (English) 7 in.
    Height (Metric) 17.7 cm
    Max. Temperature (English) 175°F
    Min. Temperature (English) -40°F
    Thickness 0.06 in.
    Width (English) 10 in.
    Width (Metric) 25.4 cm
    Temperature Range (Metric) -40°C to 80°C
    Temperature Range (English) -40°F to 175°F
    Displayed Text DANGER Hazardous Waste Storage Area Unauthorized Persons Keep Out
    Product Type Admittance Sign
    Color Red on White
    Show More Show Less

    Adhere to
    Safety Regulations and Best Practices

    Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.