Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Globally Harmonized System (GHS) Flammable Picto Labels
GSA_VA

Catalog No. p-4569857
Click to view available options
Quantity:
16/Pk.
39/Pk.
4/Pk.
Material:
Coated Paper
Vinyl
Height (English):
0.62 in.
1 in.
2 in.
Width (English):
0.6 in.
1 in.
2 in.

For flammable gases, liquids and solids; pyrophorics; self-reatives; self-heating

Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.

  • Legend: Flammable
  • Made of Vinyl or Coated paper
  • Available in three sizes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance GHS, OSHA 29 CFR 1910.1200
Color Black, Red, White
Material Vinyl
Product Type Pictogram Label
Display Text Flammable
For Use With (Application) Flammable gases, liquids and solids
Quantity 39/Pk.
Temperature Range (English) -4 to 167°F
Temperature Range (Metric) -20°C to +75°C
Width (English) 0.6 in.
Height (English) 0.62 in.
Height (Metric) 15.20 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.