Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Globally Harmonized System (GHS) Health Danger Picto Labels

Catalog No. 19140705 Shop All Brady Corporation Products
Click to view available options
Quantity:
16/Pk.
39/Pk.
4/Pk.
Material:
Paper
Vinyl
Vinyl (B-7569)

Indicates carcinogens, respiratory sensitizers, reproductive toxicity, target organ toxicity, germ cell mutagens

Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations– universal approach to classifying and communicating chemical hazards

  • Legend: Health Danger
  • Made of B-7569 viny or B-235 paper
  • Available in three sizes

Specifications

Certifications/Compliance GHS, OSHA 29 CFR 1910.1200
Adhesive Type Permanent Acrylic
Color Black, Red, White
Finish Gloss
Format Label
Material Vinyl (B-7569)
Sterility Non-sterile
For Use With (Application) For carcinogens, respiratory sensitizers, reproductive toxicity, target organ toxicity, germ cell mutagens
Quantity 4/Pk.
Dimensions (LxW) 2 x 2 in.
Width (English) 2 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.