Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Hazardous Materials Labels
Shop All Brady Corporation Products
Click to view available options
Display Text:
Acetone
Solvent N.O.S.
Description
- The B-928 is a flexible vinyl film with a cold temperature applicable pressure sensitive adhesive
- Service temperature range of -50 to 176°F
- Average outdoor durability of 2 years
- RoHS Compatibility Pass 2011/65/EU
Specifications
Specifications
Certifications/Compliance | RoHS |
Adhesive Type | Acrylic, Pressure-sensitive |
Color | Black, Yellow |
Finish | Matte |
Format | Label |
Material | Vinyl |
Product Type | Hazardous Waste Label |
Sterility | Non-sterile |
Display Text | Solvent N.O.S. |
For Use With (Application) | Hazardous waste material labels |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction