Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Key Retaining Keyed Alike Steel Padlocks with 2 inch Shackle Locks
Click to view available options
Color:
Brown
Purple
Description
Includes
1 key
- Key-retaining steel padlock has a 2 in. reinforced laminated steel shackle
- Hardened steel body withstands severe physical abuse and tampering
- Offers individually coated body plates that provide superior rust protection and corrosion resistance
- Available in a variety of different colors
Specifications
Specifications
Color | Brown |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction