Learn More
Brady™ LABXPERT™ High-Performance Lab Polyester Labels
LABXPERT™ High-Performance Lab Polyester Labels are made of self-laminating polyester (B-461), a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
Supplier:  Brady™ XSL75461
 View more versions of this product
Catalog No. 11-878-133
Description
Description
  • Self-Laminating Polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • All-in-one cartridge includes ribbon and label supply
Specifications
Specifications
Long tail
Black, White, Translucent
Label
Label
Cryo vial, Laboratory identification, Tube, Vial
1
Rectangle
-196°C to +121°C
0.048 mm
1 in.
2.6 in.
25.40 mm
Acrylic, Permanent
Matte
Polyester
Non-sterile
LabXpert™ Printer
150/Pk.
-320 to 250°F
0.0019 in.
2.625 in. H x 1 in. W
25.4 mm
66.67 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.