Learn More
Brady™ Oil Only Drag Nets
Shop All Brady Corporation Products
Click to view available options
Includes:
30 nets on 50 ft. rope
30 nets, no rope
Description
- Made up of flat, chemical-resistant filaments in half-pound units
- Can be re-used after draining or dousing
- Extracted oil can be recycled or downgraded as fuel oil
- Used as sweep between two boats or deployed along shoreline
- Available in two sizes: 30 nets on 50 ft. rope and 30 nets (no rope)
Specifications
Specifications
| Type | Oil/Drag Net |
| Includes | 30 nets, no rope |