Learn More
Brady™ Oil Only Socks
Shop All Brady Corporation Products
Click to view available options
Packaging Quantity:
12/Cs.
30/Cs.
4/Cs.
6/Cs.
Description
- Made of oil-only polypropylene SOC
- Molds around corners and conforms to uneven surfaces
- Does not absorb water or water-based solutions
- Wringable and can be incinerated
- Used as a working tool for all major industries
- Available in variety of types and sizes
Specifications
Specifications
| Material | Polypropylene |
| Type | Oil Only Socks |
| For Use With (Application) | For use against oil and petroleum based spills |
| Packaging Quantity | 6/Cs. |