Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Quick-Align™ Symbols

Catalog No. p-172688
Click to view available options
Displayed Text:
ACID
ALK
COR
No Water
OXY
Height (English):
1 in.
2 in.
3 in.
4 in.

For use with Blank Hazardous Communication Panels to identify hazardous materials in accordance with the NFPA system

  • Tough outdoor-grade self-sticking vinyl characters are black Gothic style (except for radiation trefoil)
  • Speciific hazard warnings (“Use no water”, radiation symbol, oxidizer, acid, alkalai, corrosive)

Always consult Material Safety Data Sheets for product-specific hazard information.

Order Info

Each package contains 10 identical characters.

Specifications

Certifications/Compliance RoHS
Material Vinyl
Quantity 10/Pk.
Style Gothic
For Use With (Application) Indoor and outdoor use, used for pipemarkers, arrow tape, numbers and letters, vehicle placards, pedestal markers, warning panels and safety signs
Height (English) 4 in.
Max. Temperature (English) 180°F
Min. Temperature (English) -40°F
Thickness 0.004 in.
Temperature Range (Metric) -40°C to 82°C
Temperature Range (English) -40°F to 180°F
Displayed Text ACID
Product Type Hazardous Material Identifier
Color Black
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.